missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Bis(4-bromophenyl)-2-propanone 99.0+%, TCI America™
Supplier: TCI America B47911G
Specifications
| 1,3-Bis(4-bromophenyl)-2-propanone | |
| 119°C | |
| C15H12Br2O | |
| 1 g | |
| PQDQAUVBWSUMMB-UHFFFAOYSA-N | |
| 1,3-bis(4-bromophenyl)propan-1-one | |
| 603327 | |
| ≥99.0% (GC) |
| 54523-47-6 | |
| White-Yellow | |
| MFCD03843365 | |
| 4,4′C-Dibromodibenzyl Ketone, 1,3-Bis(4-bromophenyl)acetone | |
| C1=CC(=CC=C1CCC(=O)C2=CC=C(C=C2)Br)Br | |
| 368.068 | |
| 368.07 | |
| Crystalline Powder |
Chemical Identifiers
| 54523-47-6 | |
| 368.068 | |
| PQDQAUVBWSUMMB-UHFFFAOYSA-N | |
| 603327 | |
| C1=CC(=CC=C1CCC(=O)C2=CC=C(C=C2)Br)Br |
| C15H12Br2O | |
| MFCD03843365 | |
| 4,4′C-Dibromodibenzyl Ketone, 1,3-Bis(4-bromophenyl)acetone | |
| 1,3-bis(4-bromophenyl)propan-1-one |
Safety and Handling
TSCA : No