missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(4-bromophenyl)-2-propanone 99.0+%, TCI America™

Catalog No. p-7123752
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4791200MG 200 mg
B47911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4791200MG Supplier TCI America Supplier No. B4791200MG

Chemical Identifiers

CAS 54523-47-6
Molecular Formula C15H12Br2O
Molecular Weight (g/mol) 368.068
MDL Number MFCD03843365
InChI Key PQDQAUVBWSUMMB-UHFFFAOYSA-N
Synonym 4,4′C-Dibromodibenzyl Ketone, 1,3-Bis(4-bromophenyl)acetone
PubChem CID 603327
IUPAC Name 1,3-bis(4-bromophenyl)propan-1-one
SMILES C1=CC(=CC=C1CCC(=O)C2=CC=C(C=C2)Br)Br

Specifications

Melting Point 119°C
Color White-Yellow
Quantity 200 mg
Formula Weight 368.07
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(4-bromophenyl)-2-propanone