missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(4-aminophenoxy)benzene 98.0+%, TCI America™

Catalog No. b168025g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B168025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B168025G Supplier TCI America Supplier No. B168025G
Only null left

Chemical Identifiers

CAS 2479-46-1
Molecular Formula C18H16N2O2
Molecular Weight (g/mol) 292.338
MDL Number MFCD00039154
InChI Key WUPRYUDHUFLKFL-UHFFFAOYSA-N
Synonym Resorcinol Bis(4-aminophenyl) Ether
PubChem CID 17201
IUPAC Name 4-[3-(4-aminophenoxy)phenoxy]aniline
SMILES C1=CC(=CC(=C1)OC2=CC=C(C=C2)N)OC3=CC=C(C=C3)N

Specifications

Melting Point 116°C
Quantity 25 g
Formula Weight 292.34
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(4-aminophenoxy)benzene
EINECSNumber (3)-4173
RTECSNumber BY8236000
TSCA Yes