accessibility menu, dialog, popup

UserName

NH2OOH2N

CAS RN 2479-46-1

IUPAC Name: 4-[3-(4-aminophenoxy)phenoxy]aniline
Synonyms: 4-[3-(4-aminophenoxy)phenoxy]aniline
Molecular Weight (g/mol): 292.34
Molecular Formula: C18H16N2O2
InChi Key: WUPRYUDHUFLKFL-UHFFFAOYSA-N
SMILES: NC1=CC=C(OC2=CC(OC3=CC=C(N)C=C3)=CC=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results