missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Bis(3-hydroxy-3-pentyl)benzene 90.0+%, TCI America™
Supplier: TCI America B343625G
Specifications
| 1,3-Bis(3-hydroxy-3-pentyl)benzene | |
| White-Yellow | |
| 25 g | |
| DBVBDLSFXMYKIU-UHFFFAOYSA-N | |
| 3-[3-(3-hydroxypentan-3-yl)phenyl]pentan-3-ol | |
| 44630004 | |
| ≥90.0% (GC) |
| 676465-94-4 | |
| C16H26O2 | |
| 1,3-Bis(3-hydroxy-3-amyl)benzene, 1,3-Bis(1-ethyl-1-hydroxypropyl)benzene, 3,3′-(m-Phenylene)di-3-pentanol, alpha,alpha,alpha′,alpha′-Tetraethyl-1,3-benzenedimethanol | |
| CCC(CC)(C1=CC(=CC=C1)C(CC)(CC)O)O | |
| 250.382 | |
| 250.38 | |
| Crystalline Powder |
Chemical Identifiers
| 676465-94-4 | |
| 250.382 | |
| 44630004 | |
| CCC(CC)(C1=CC(=CC=C1)C(CC)(CC)O)O |
| C16H26O2 | |
| DBVBDLSFXMYKIU-UHFFFAOYSA-N | |
| 3-[3-(3-hydroxypentan-3-yl)phenyl]pentan-3-ol |
Safety and Handling
TSCA : No