missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(3-hydroxy-3-pentyl)benzene 90.0+%, TCI America™

Catalog No. B34365G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34365G 5 g
B343625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34365G Supplier TCI America Supplier No. B34365G
Only null left

Chemical Identifiers

CAS 676465-94-4
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.382
InChI Key DBVBDLSFXMYKIU-UHFFFAOYSA-N
PubChem CID 44630004
IUPAC Name 3-[3-(3-hydroxypentan-3-yl)phenyl]pentan-3-ol
SMILES CCC(CC)(C1=CC(=CC=C1)C(CC)(CC)O)O

Specifications

Color White-Yellow
Quantity 5 g
Synonym 1,3-Bis(3-hydroxy-3-amyl)benzene, 1,3-Bis(1-ethyl-1-hydroxypropyl)benzene, 3,3′C-(m-Phenylene)di-3-pentanol, alpha,alpha,alpha′C,alpha′C-Tetraethyl-1,3-benzenedimethanol
Formula Weight 250.38
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(3-hydroxy-3-pentyl)benzene