missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Bis(3,3,3-trifluoropropyl)-1,1,3,3-tetramethyldisilazane 94.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 119°C |
| Quantity | 5 g |
| Formula Weight | 325.45 |
| Percent Purity | ≥94.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1,3-Bis(3,3,3-trifluoropropyl)-1,1,3,3-tetramethyldisilazane |
Safety and Handling
| TSCA | Yes |