missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Bis(2-propynyloxy)benzene 97.0+%, TCI America™
$225.56 - $781.03
Chemical Identifiers
| CAS | 26627-36-1 |
|---|---|
| Molecular Formula | C12H10O2 |
| Molecular Weight (g/mol) | 186.21 |
| MDL Number | MFCD00224312 |
| InChI Key | HSRDPYWXTZGWFQ-UHFFFAOYSA-N |
| Synonym | 1,3-Bis(propargyloxy)benzene |
| PubChem CID | 11030523 |
| IUPAC Name | 1,3-bis(prop-2-ynoxy)benzene |
| SMILES | C#CCOC1=CC(=CC=C1)OCC#C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B4521200MG
|
TCI America
B4521200MG |
200 mg |
Each for $225.56
|
|
|||||
|
B45211G
|
TCI America
B45211G |
1 g |
Each for $781.03
|
|
|||||
Chemical Identifiers
| 26627-36-1 | |
| 186.21 | |
| HSRDPYWXTZGWFQ-UHFFFAOYSA-N | |
| 11030523 | |
| C#CCOC1=CC(=CC=C1)OCC#C |
| C12H10O2 | |
| MFCD00224312 | |
| 1,3-Bis(propargyloxy)benzene | |
| 1,3-bis(prop-2-ynoxy)benzene |
Specifications
| 26627-36-1 | |
| White-Yellow | |
| MFCD00224312 | |
| 1,3-Bis(propargyloxy)benzene | |
| C#CCOC1=CC(=CC=C1)OCC#C | |
| 186.21 | |
| 186.21 | |
| Crystalline Powder |
| 40°C | |
| C12H10O2 | |
| 200 mg | |
| HSRDPYWXTZGWFQ-UHFFFAOYSA-N | |
| 1,3-bis(prop-2-ynoxy)benzene | |
| 11030523 | |
| ≥97.0% (GC) | |
| 1,3-Bis(2-propynyloxy)benzene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator