missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Bis(2-propynyloxy)benzene 97.0+%, TCI America™
Supplier: TCI America B4521200MG
Specifications
| 1,3-Bis(2-propynyloxy)benzene | |
| 40°C | |
| C12H10O2 | |
| 200 mg | |
| HSRDPYWXTZGWFQ-UHFFFAOYSA-N | |
| 1,3-bis(prop-2-ynoxy)benzene | |
| 11030523 | |
| ≥97.0% (GC) |
| 26627-36-1 | |
| White-Yellow | |
| MFCD00224312 | |
| 1,3-Bis(propargyloxy)benzene | |
| C#CCOC1=CC(=CC=C1)OCC#C | |
| 186.21 | |
| 186.21 | |
| Crystalline Powder |
Chemical Identifiers
| 26627-36-1 | |
| 186.21 | |
| HSRDPYWXTZGWFQ-UHFFFAOYSA-N | |
| 11030523 | |
| C#CCOC1=CC(=CC=C1)OCC#C |
| C12H10O2 | |
| MFCD00224312 | |
| 1,3-Bis(propargyloxy)benzene | |
| 1,3-bis(prop-2-ynoxy)benzene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator