missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(2-hydroxyethoxy)benzene 98.0+%, TCI America™

Catalog No. p-7119167
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B162525G 25 g
B1625500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B162525G Supplier TCI America Supplier No. B162525G

Chemical Identifiers

CAS 102-40-9
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.22
MDL Number MFCD00016566
InChI Key IAXFZZHBFXRZMT-UHFFFAOYSA-N
Synonym Resorcinol Bis(beta-hydroxyethyl) Ether
PubChem CID 66885
IUPAC Name 2-[3-(2-hydroxyethoxy)phenoxy]ethan-1-ol
SMILES OCCOC1=CC(OCCO)=CC=C1

Specifications

Melting Point 90°C
Color White
Boiling Point 234°C
Quantity 25 g
Formula Weight 198.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(2-hydroxyethoxy)benzene
EINECSNumber (3)-3864
TSCA Yes