missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(2,6-diisopropylphenyl)imidazolinium Chloride 96.0+%, TCI America™

Catalog No. B31571G
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
500 mg
Catalog No. Quantity
B31571G 1 g
B3157500MG 500 mg
B31575G 5 g
3 options

Catalog No. B31571G

Supplier: TCI America B31571G

Only null left

Chemical Identifiers

CAS 258278-25-0
Molecular Formula C27H39ClN2
Molecular Weight (g/mol) 427.07
MDL Number MFCD07369796
InChI Key LWPXTYZKAWSRIP-UHFFFAOYSA-M
Synonym 1,3-bis 2,6-diisopropylphenyl-4,5-dihydro-1h-imidazol-3-ium chloride, sipr.hcl, 1,3-bis 2,6-diisopropylphenyl-imidazolidinium-chloride, 1,3-bis 2,6-diisopropylphenyl imidazolidinium chloride, n,n'-2,6-diisopropylphenyl dihydroimidazolium chloride, 1,3-bis 2,6-diisopropylphenyl-4,5-dihydro-1??-imidazol-1-ylium chloride, 1,3-bis 2,6-diisopropylphenyl imidazolinium chloride?, 1,3-bis-2,6-diisopropylphenyl imidazolidinium chloride, 1,3-bis-2,6-diisopropylphenyl imidazolidiniumchloride, 1,3-bis 2,6-bis isopropyl phenyl imidazolin-3-ium chloride
PubChem CID 2734918
IUPAC Name 1,3-bis[2,6-bis(propan-2-yl)phenyl]-4,5-dihydro-3H-1λ⁵-imidazol-1-ylium chloride
SMILES [Cl-].CC(C)C1=CC=CC(C(C)C)=C1N1CC[N+](=C1)C1=C(C=CC=C1C(C)C)C(C)C

Specifications

Melting Point 293°C
Color White-Yellow
Quantity 1 g
Formula Weight 427.07
Percent Purity ≥96.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(2,6-diisopropylphenyl)imidazolinium Chloride