missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene 98.0+%, TCI America™

Catalog No. B34651G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34651G 1 g
B34655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34651G Supplier TCI America Supplier No. B34651G

Chemical Identifiers

CAS 244187-81-3
Molecular Formula C27H36N2
Molecular Weight (g/mol) 388.599
MDL Number MFCD07781991
InChI Key VYCIHDBIKGRENI-UHFFFAOYSA-N
Synonym IPr
PubChem CID 11271730
IUPAC Name 1,3-bis[2,6-di(propan-2-yl)phenyl]-2H-imidazol-1-ium-2-ide
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N2C=C[N+](=[C-]2)C3=C(C=CC=C3C(C)C)C(C)C

Specifications

Melting Point 217°C
Color White-Yellow
Quantity 1 g
Formula Weight 388.60
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene
TSCA No
Recommended Storage Refrigerator