CAS RN 244187-81-3
IUPAC Name:
1,3-bis[2,6-bis(propan-2-yl)phenyl]-3H-1λ⁵-imidazol-1-ylium-2-ide
Synonyms:
1,3-bis(2,6-diisopropylphenyl)-1λ⁵-imidazol-1-ylium-2-ide
Molecular Weight (g/mol):
388.6
Molecular Formula:
C27H36N2
InChi Key:
VYCIHDBIKGRENI-UHFFFAOYSA-N
SMILES:
CC(C)C1=CC=CC(C(C)C)=C1N1C=C[N+](=[C-]1)C1=C(C=CC=C1C(C)C)C(C)C
1
–
1
of
1
results
1
1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene 98.0+%, TCI America™
CAS: 244187-81-3 Molecular Formula: C27H36N2 Molecular Weight (g/mol): 388.599 MDL Number: MFCD07781991 InChI Key: VYCIHDBIKGRENI-UHFFFAOYSA-N Synonym: IPr PubChem CID: 11271730 IUPAC Name: 1,3-bis[2,6-di(propan-2-yl)phenyl]-2H-imidazol-1-ium-2-ide SMILES: CC(C)C1=C(C(=CC=C1)C(C)C)N2C=C[N+](=[C-]2)C3=C(C=CC=C3C(C)C)C(C)C