missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene](1,3-diphenyl-1,3-propanedionato)copper(I) 98.0+%, TCI America™
$109.98 - $326.21
Chemical Identifiers
| CAS | 920739-11-3 |
|---|---|
| Molecular Formula | C42H48CuN2O2 |
| Molecular Weight (g/mol) | 676.40 |
| MDL Number | MFCD12545959 |
| InChI Key | QEXNUQSCWMWURE-UHFFFAOYSA-N |
| PubChem CID | 91659139 |
| IUPAC Name | 1,3-bis[2,6-bis(propan-2-yl)phenyl]-2,3-dihydro-1H-imidazol-2-ylidene 3-hydroxy-1,3-diphenylprop-2-en-1-one copper |
| SMILES | [Cu].OC(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1.CC(C)C1=CC=CC(C(C)C)=C1N1[C]N(C=C1)C1=C(C=CC=C1C(C)C)C(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B3351200MG
|
TCI America
B3351200MG |
200 mg |
Each for $109.98
|
|
|||||
|
B33511G
|
TCI America
B33511G |
1 g |
Each for $326.21
|
|
|||||
Chemical Identifiers
| 920739-11-3 | |
| 676.40 | |
| QEXNUQSCWMWURE-UHFFFAOYSA-N | |
| 1,3-bis[2,6-bis(propan-2-yl)phenyl]-2,3-dihydro-1H-imidazol-2-ylidene 3-hydroxy-1,3-diphenylprop-2-en-1-one copper |
| C42H48CuN2O2 | |
| MFCD12545959 | |
| 91659139 | |
| [Cu].OC(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1.CC(C)C1=CC=CC(C(C)C)=C1N1[C]N(C=C1)C1=C(C=CC=C1C(C)C)C(C)C |
Specifications
| 920739-11-3 | |
| MFCD12545959 | |
| QEXNUQSCWMWURE-UHFFFAOYSA-N | |
| 1,3-bis[2,6-bis(propan-2-yl)phenyl]-2,3-dihydro-1H-imidazol-2-ylidene 3-hydroxy-1,3-diphenylprop-2-en-1-one copper | |
| 91659139 | |
| ≥98.0% (T) | |
| [1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene](1,3-diphenyl-1,3-propanedionato)copper(I) |
| C42H48CuN2O2 | |
| 200 mg | |
| [Cu].OC(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1.CC(C)C1=CC=CC(C(C)C)=C1N1[C]N(C=C1)C1=C(C=CC=C1C(C)C)C(C)C | |
| 676.40 | |
| 675.40 | |
| Crystalline Powder |
Safety and Handling
TSCA : No