missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene](1,3-diphenyl-1,3-propanedionato)copper(I) 98.0+%, TCI America™
Supplier: TCI America B33511G
Specifications
| [1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene](1,3-diphenyl-1,3-propanedionato)copper(I) | |
| C42H48CuN2O2 | |
| 1 g | |
| [Cu].OC(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1.CC(C)C1=CC=CC(C(C)C)=C1N1[C]N(C=C1)C1=C(C=CC=C1C(C)C)C(C)C | |
| 676.40 | |
| 675.40 | |
| Crystalline Powder |
| 920739-11-3 | |
| MFCD12545959 | |
| QEXNUQSCWMWURE-UHFFFAOYSA-N | |
| 1,3-bis[2,6-bis(propan-2-yl)phenyl]-2,3-dihydro-1H-imidazol-2-ylidene 3-hydroxy-1,3-diphenylprop-2-en-1-one copper | |
| 91659139 | |
| ≥98.0% (T) |
Chemical Identifiers
| 920739-11-3 | |
| 676.40 | |
| QEXNUQSCWMWURE-UHFFFAOYSA-N | |
| 1,3-bis[2,6-bis(propan-2-yl)phenyl]-2,3-dihydro-1H-imidazol-2-ylidene 3-hydroxy-1,3-diphenylprop-2-en-1-one copper |
| C42H48CuN2O2 | |
| MFCD12545959 | |
| 91659139 | |
| [Cu].OC(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1.CC(C)C1=CC=CC(C(C)C)=C1N1[C]N(C=C1)C1=C(C=CC=C1C(C)C)C(C)C |
Safety and Handling
TSCA : No