missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis[(2,2-dimethyl-1,3-dioxan-5-yl)oxy]-2-propanol 98.0+%, TCI America™

Catalog No. B3905100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3905100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B3905100MG Supplier TCI America Supplier No. B3905100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 847682-00-2
Molecular Formula C15H28O7
Molecular Weight (g/mol) 320.382
InChI Key LKMPEWGOAMMQKB-UHFFFAOYSA-N
PubChem CID 49765605
IUPAC Name 1,3-bis[(2,2-dimethyl-1,3-dioxan-5-yl)oxy]propan-2-ol
SMILES CC1(OCC(CO1)OCC(COC2COC(OC2)(C)C)O)C

Specifications

Melting Point 55°C
Color White
Quantity 100 mg
Formula Weight 320.38
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis[(2,2-dimethyl-1,3-dioxan-5-yl)oxy]-2-propanol
TSCA No
Recommended Storage Refrigerator