accessibility menu, dialog, popup

UserName

H3COOOOOOCH3CH3OHH3C

CAS RN 847682-00-2

IUPAC Name: 1,3-bis[(2,2-dimethyl-1,3-dioxan-5-yl)oxy]propan-2-ol
Synonyms: 1,3-bis[(2,2-dimethyl-1,3-dioxan-5-yl)oxy]propan-2-ol
Molecular Weight (g/mol): 320.38
Molecular Formula: C15H28O7
InChi Key: LKMPEWGOAMMQKB-UHFFFAOYSA-N
SMILES: CC1(C)OCC(CO1)OCC(O)COC1COC(C)(C)OC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results