missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzothiazole-6-carboxaldehyde, 97%

Catalog No. AC431150010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC431150010 1 g Glass bottle
AC431150050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC431150010 Supplier Thermo Scientific Chemicals Supplier No. 431150010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 19989-67-4 | C8H5NOS | 163.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19989-67-4
Molecular Formula C8H5NOS
Molecular Weight (g/mol) 163.2
InChI Key AVSFPLJXSHRMHM-UHFFFAOYSA-N
Synonym 6-benzothiazolecarboxaldehyde, benzo d thiazole-6-carbaldehyde, 1,3-benzothiazole-6-carboxaldehyde, 6-formylbenzothiazole, benzothiazol-6-carboxaldehyde, 6-benzothiazolecarbaldehyde, 1?3-benzothiazole-6-carboxaldehyde, 6-benzo d thiazolecarboxaldehyde, 6-benzothiazolecarboxaldehyde 8ci,9ci
PubChem CID 15089709
IUPAC Name 1,3-benzothiazole-6-carbaldehyde
SMILES C1=CC2=C(C=C1C=O)SC=N2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 92.0°C to 93.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 163.2
Percent Purity 97%
Physical Form Crystalline Powder or Low Melting Solid
Chemical Name or Material 1, 3-Benzothiazole-6-carboxaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only