CAS RN 19989-67-4
IUPAC Name:
1,3-benzothiazole-6-carbaldehyde
Synonyms:
1,3-benzothiazole-6-carbaldehyde
Molecular Weight (g/mol):
163.19
Molecular Formula:
C8H5NOS
InChi Key:
AVSFPLJXSHRMHM-UHFFFAOYSA-N
SMILES:
O=CC1=CC=C2N=CSC2=C1
1
–
1
of
1
results
1
Promotions Available
1,3-Benzothiazole-6-carboxaldehyde, 97%
1,3-Benzothiazole-6-carboxaldehyde, 97%
CAS: 19989-67-4 Molecular Formula: C8H5NOS Molecular Weight (g/mol): 163.2 InChI Key: AVSFPLJXSHRMHM-UHFFFAOYSA-N Synonym: 6-benzothiazolecarboxaldehyde,benzo d thiazole-6-carbaldehyde,1,3-benzothiazole-6-carboxaldehyde,6-formylbenzothiazole,benzothiazol-6-carboxaldehyde,6-benzothiazolecarbaldehyde,1?3-benzothiazole-6-carboxaldehyde,6-benzo d thiazolecarboxaldehyde,6-benzothiazolecarboxaldehyde 8ci,9ci PubChem CID: 15089709 IUPAC Name: 1,3-benzothiazole-6-carbaldehyde SMILES: C1=CC2=C(C=C1C=O)SC=N2