missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzothiazole-2-carbaldehyde, Thermo Scientific™

Catalog No. p-2694682
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC06004DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC06004DA Supplier Thermo Scientific Chemicals Supplier No. CC06004DA
Only null left

Chemical Identifiers

CAS 6639-57-2
Molecular Formula C8H5NOS
Molecular Weight (g/mol) 163.194
MDL Number MFCD00526215
InChI Key RHKPJTFLRQNNGJ-UHFFFAOYSA-N
Synonym 2-benzothiazolecarboxaldehyde, benzothiazole-2-carbaldehyde, benzothiazole-2-carboxaldehyde, benzo d thiazole-2-carbaldehyde, 1,3-benzothiazole-2-carboxaldehyde, 2-formylbenzothiazole, zlchem 104, benzothiazolecarbaldehyde, pubchem14482, 2-benzothiazolecarbaldehyde
PubChem CID 241608
IUPAC Name 1,3-benzothiazole-2-carbaldehyde
SMILES C1=CC=C2C(=C1)N=C(S2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 163.2
Percent Purity 97%
Chemical Name or Material 1, 3-Benzothiazole-2-carbaldehyde