accessibility menu, dialog, popup

UserName

CAS RN 6639-57-2

IUPAC Name: 1,3-benzothiazole-2-carbaldehyde
Synonyms: 1,3-benzothiazole-2-carbaldehyde
Molecular Weight (g/mol): 163.19
Molecular Formula: C8H5NOS
InChi Key: RHKPJTFLRQNNGJ-UHFFFAOYSA-N
SMILES: O=CC1=NC2=CC=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results