missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzothiazol-6-amine, 97+%, Thermo Scientific™

Catalog No. p-4509893
Change view
Click to view available options
Quantity:
1g
25g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC05914DA 1g Amber Glass Bottle
CC05914EB 25g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC05914DA Supplier Thermo Scientific Chemicals Supplier No. CC05914DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 533-30-2
Molecular Formula C7H6N2S
Molecular Weight (g/mol) 150.199
InChI Key FAYAYUOZWYJNBD-UHFFFAOYSA-N
Synonym 6-aminobenzothiazole, 6-benzothiazolamine, benzo d thiazol-6-amine, 6-amino-benzothiazole, benzothiazole, 6-amino, benzothiazol-6-amine, benzothiazol-6-ylamine, unii-9yfg4wuj1g, 9yfg4wuj1g, 1,3-benzothiazol-6-ylamine
PubChem CID 68288
IUPAC Name 1,3-benzothiazol-6-amine
SMILES C1=CC2=C(C=C1N)SC=N2

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Merck Index 15, 420
Formula Weight 150.2
Percent Purity 97%
Chemical Name or Material 1, 3-Benzothiazol-6-amine