missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzenedithiol 95.0+%, TCI America™

Catalog No. B43715G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43715G 5 g
B43711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43715G Supplier TCI America Supplier No. B43715G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 626-04-0
Molecular Formula C6H6S2
Molecular Weight (g/mol) 142.23
MDL Number MFCD00004842
InChI Key ZWOASCVFHSYHOB-UHFFFAOYSA-N
Synonym 1,3-benzenedithiol, 1,3-dimercaptobenzene, dithioresorcin, dithioresorcinol, 1,3-benzedithiol, acmc-20ap5g, ksc357g0j
PubChem CID 522062
IUPAC Name benzene-1,3-dithiol
SMILES SC1=CC(S)=CC=C1

Specifications

Melting Point 27°C
Color White
Boiling Point 245°C
Quantity 5 g
Formula Weight 142.23
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,3-Benzenedithiol