missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzenedimethanol, 98%

Catalog No. p-7026996
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2341703 1 g
AAB2341706 5 g
AAB2341714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2341703 Supplier Thermo Scientific Chemicals Supplier No. B2341703
Only null left

CAS: 626-18-6 | C8H10O2 | 138.166 g/mol

1,3-Benzenedimethanol was used in the synthesis of a series of cross-linked poly(orthocarbonate)s. It was also used in the synthesis of mixed- tethered system, required for the preparation of e-edge-[60]fullerenylmethanodihydropyrrole adduct.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Benzenedimethanol was used in the synthesis of a series of cross-linked poly(orthocarbonate)s. It was also used in the synthesis of mixed- tethered system, required for the preparation of e-edge-[60]fullerenylmethanodihydropyrrole adduct.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agent.

Chemical Identifiers

CAS 626-18-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00004649
InChI Key YWMLORGQOFONNT-UHFFFAOYSA-N
Synonym 1,3-benzenedimethanol, 3-hydroxymethyl phenyl methanol, 1,3-phenylenedimethanol, 1,3-dihydroxymethylbenzene, m-xylene-alpha,alpha'-diol, m-xylene glycol, m-xylylene glycol, 3-hydroxymethyl phenyl methan-1-ol, m-xylylenalkohol, m-benzenedimethanol
PubChem CID 69374
IUPAC Name [3-(hydroxymethyl)phenyl]methanol
SMILES C1=CC(=CC(=C1)CO)CO

Specifications

Melting Point 56°C to 60°C
Boiling Point 154°C to 159°C (13 mmHg)
Quantity 1 g
Beilstein 1562398
Solubility Information Soluble in water.
Formula Weight 138.17
Percent Purity 98%
Chemical Name or Material 1,3-Benzenedimethanol
Hazard Category H302-H319
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
EINECSNumber 210-934-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only