missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Aminopropyl)imidazole 97.0+%, TCI America™

Catalog No. p-7117296
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A118525G 25 g
A1185100G 100 g
A1185500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A118525G Supplier TCI America Supplier No. A118525G
Only null left

Chemical Identifiers

CAS 5036-48-6
Molecular Formula C6H11N3
Molecular Weight (g/mol) 125.18
MDL Number MFCD00009819
InChI Key KDHWOCLBMVSZPG-UHFFFAOYSA-N
Synonym 1-3-aminopropyl imidazole, n-3-aminopropyl imidazole, 1h-imidazole-1-propanamine, n-3-aminopropyl-imidazole, 3-1h-imidazol-1-yl propan-1-amine, 3-imidazol-1-ylpropylamine, 3-1h-imidazol-1-yl-1-propanamine, 1-3-aminopropyl-imidazole, 3-imidazol-1-yl propylamine, 3-imidazolylpropylamine
PubChem CID 78736
IUPAC Name 3-(1H-imidazol-1-yl)propan-1-amine
SMILES NCCCN1C=CN=C1

Specifications

Color Yellow
Boiling Point 117°C
Quantity 25 g
UN Number 2735
Formula Weight 125.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(3-Aminopropyl)imidazole