accessibility menu, dialog, popup

UserName

H2NNN

CAS RN 5036-48-6

IUPAC Name: 3-(1H-imidazol-1-yl)propan-1-amine
Synonyms: 3-(imidazol-1-yl)propan-1-amine
Molecular Weight (g/mol): 125.18
Molecular Formula: C6H11N3
InChi Key: KDHWOCLBMVSZPG-UHFFFAOYSA-N
SMILES: NCCCN1C=CN=C1
Boiling Point 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)