missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Aminopropyl)imidazole, 98%

Catalog No. AAA1416918
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1416918 50 g
AAA1416930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1416918 Supplier Thermo Scientific Chemicals Supplier No. A1416918
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5036-48-6 | C6H11N3 | 125.18 g/mol

1-(3-Aminopropyl)imidazole is used in the synthesis of pH-sensitive polyaspartamide derivatives. It is also used in the preparation of pH-sensitive amphiphilic polymers with different degrees of octaceylamine (C18) substitution by graft reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(3-Aminopropyl)imidazole is used in the synthesis of pH-sensitive polyaspartamide derivatives. It is also used in the preparation of pH-sensitive amphiphilic polymers with different degrees of octaceylamine (C18) substitution by graft reaction.

Solubility
Fully miscible in water.

Notes
Store away from oxidizing agents and air. Air sensitive. Store under dry inert gas. Incompatible with air and carbondioxide.

Chemical Identifiers

CAS 5036-48-6
Molecular Formula C6H11N3
Molecular Weight (g/mol) 125.18
MDL Number MFCD00009819
InChI Key KDHWOCLBMVSZPG-UHFFFAOYSA-N
Synonym 1-3-aminopropyl imidazole, n-3-aminopropyl imidazole, 1h-imidazole-1-propanamine, n-3-aminopropyl-imidazole, 3-1h-imidazol-1-yl propan-1-amine, 3-imidazol-1-ylpropylamine, 3-1h-imidazol-1-yl-1-propanamine, 1-3-aminopropyl-imidazole, 3-imidazol-1-yl propylamine, 3-imidazolylpropylamine
PubChem CID 78736
SMILES NCCCN1C=CN=C1

Specifications

Melting Point -68°C
Density 1.062
pH 11.3
Boiling Point 296°C
Flash Point 154°C (309°F)
Odor Amine-like
Refractive Index 1.519
Quantity 50 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
IUPAC Name 3-(1H-imidazol-1-yl)propan-1-amine
Formula Weight 125.18
Percent Purity 98%
Chemical Name or Material 1-(3-Aminopropyl)imidazole
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 225-730-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only