Learn More
1-(3-Aminophenyl)ethanol, 98%
CAS: 2454-37-7 | C8H11NO | 137.182 g/mol
Supplier: Thermo Scientific Chemicals L0651314
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1-(3-Aminophenyl)ethanol | |
| 66°C to 69°C | |
| 157°C (314°F) | |
| MFCD00007798 | |
| 2802415 | |
| QPKNDHZQPGMLCJ-UHFFFAOYSA-N | |
| 1-(3-aminophenyl)ethanol | |
| 222461 | |
| 98% |
| 2454-37-7 | |
| 217°C | |
| C8H11NO | |
| 25 g | |
| 1-3-aminophenyl ethanol, 3-1-hydroxyethyl aniline, m-1-hydroxyethyl aniline, m-amino-alpha-methylbenzyl alcohol, m-aminophenyl methyl carbinol, 1-3-aminophenyl ethan-1-ol, 3-amino-alpha-methylbenzyl alcohol, benzyl alcohol, m-amino-alpha-methyl, 3-aminophenyl-1-ethanol, benzyl alcohol, m-amino-.alpha.-methyl | |
| CC(C1=CC(=CC=C1)N)O | |
| 137.182 | |
| 137.18 |
Chemical Identifiers
| 2454-37-7 | |
| 137.182 | |
| QPKNDHZQPGMLCJ-UHFFFAOYSA-N | |
| 222461 | |
| CC(C1=CC(=CC=C1)N)O |
| C8H11NO | |
| MFCD00007798 | |
| 1-3-aminophenyl ethanol, 3-1-hydroxyethyl aniline, m-1-hydroxyethyl aniline, m-amino-alpha-methylbenzyl alcohol, m-aminophenyl methyl carbinol, 1-3-aminophenyl ethan-1-ol, 3-amino-alpha-methylbenzyl alcohol, benzyl alcohol, m-amino-alpha-methyl, 3-aminophenyl-1-ethanol, benzyl alcohol, m-amino-.alpha.-methyl | |
| 1-(3-aminophenyl)ethanol |
Safety and Handling
EINECSNumber : 219-525-3
RTECSNumber : DN5600000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only