missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Aminophenyl)ethanol, 98%

Catalog No. p-7044455
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0651314 25 g
AAL0651322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0651314 Supplier Thermo Scientific Chemicals Supplier No. L0651314

CAS: 2454-37-7 | C8H11NO | 137.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2454-37-7
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00007798
InChI Key QPKNDHZQPGMLCJ-UHFFFAOYSA-N
Synonym 1-3-aminophenyl ethanol, 3-1-hydroxyethyl aniline, m-1-hydroxyethyl aniline, m-amino-alpha-methylbenzyl alcohol, m-aminophenyl methyl carbinol, 1-3-aminophenyl ethan-1-ol, 3-amino-alpha-methylbenzyl alcohol, benzyl alcohol, m-amino-alpha-methyl, 3-aminophenyl-1-ethanol, benzyl alcohol, m-amino-.alpha.-methyl
PubChem CID 222461
IUPAC Name 1-(3-aminophenyl)ethanol
SMILES CC(C1=CC(=CC=C1)N)O

Specifications

Melting Point 66°C to 69°C
Boiling Point 217°C
Flash Point 157°C (314°F)
Quantity 25 g
Beilstein 2802415
Formula Weight 137.18
Percent Purity 98%
Chemical Name or Material 1-(3-Aminophenyl)ethanol
EINECSNumber 219-525-3
RTECSNumber DN5600000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only