missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Acetonedicarboxylic Acid 95.0+%, TCI America™
$104.38 - $939.48
Chemical Identifiers
| CAS | 542-05-2 |
|---|---|
| Molecular Formula | C5H6O5 |
| Molecular Weight (g/mol) | 146.098 |
| MDL Number | MFCD00002711 |
| InChI Key | OXTNCQMOKLOUAM-UHFFFAOYSA-N |
| Synonym | 1,3-acetonedicarboxylic acid, 3-oxoglutaric acid, acetonedicarboxylic acid, pentanedioic acid, 3-oxo, acetone-1,3-dicarboxylic acid, 3-ketoglutaric acid, unii-ih7p7wo21p, 1,3-acetonedicarboxylic acid; 3-ketoglutaric acid, ih7p7wo21p, acetondicarbonsaure |
| PubChem CID | 68328 |
| IUPAC Name | 3-oxopentanedioic acid |
| SMILES | C(C(=O)CC(=O)O)C(=O)O |
Chemical Identifiers
| 542-05-2 | |
| 146.098 | |
| OXTNCQMOKLOUAM-UHFFFAOYSA-N | |
| 68328 | |
| C(C(=O)CC(=O)O)C(=O)O |
| C5H6O5 | |
| MFCD00002711 | |
| 1,3-acetonedicarboxylic acid, 3-oxoglutaric acid, acetonedicarboxylic acid, pentanedioic acid, 3-oxo, acetone-1,3-dicarboxylic acid, 3-ketoglutaric acid, unii-ih7p7wo21p, 1,3-acetonedicarboxylic acid; 3-ketoglutaric acid, ih7p7wo21p, acetondicarbonsaure | |
| 3-oxopentanedioic acid |
Specifications
| 542-05-2 | |
| C5H6O5 | |
| 25 g | |
| OXTNCQMOKLOUAM-UHFFFAOYSA-N | |
| 3-oxopentanedioic acid | |
| 68328 | |
| ≥95.0% (T) | |
| 1,3-Acetonedicarboxylic Acid |
| 126°C | |
| MFCD00002711 | |
| 1,3-acetonedicarboxylic acid, 3-oxoglutaric acid, acetonedicarboxylic acid, pentanedioic acid, 3-oxo, acetone-1,3-dicarboxylic acid, 3-ketoglutaric acid, unii-ih7p7wo21p, 1,3-acetonedicarboxylic acid; 3-ketoglutaric acid, ih7p7wo21p, acetondicarbonsaure | |
| C(C(=O)CC(=O)O)C(=O)O | |
| 146.098 | |
| 146.10 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-2762
TSCA : Yes
Recommended Storage : Freezer