missing translation for 'onlineSavingsMsg'
Learn More

1,3-Acetonedicarboxylic Acid 95.0+%, TCI America™

Catalog No. K000625G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K000625G 25 g
K0006250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K000625G Supplier TCI America Supplier No. K000625G

Chemical Identifiers

CAS 542-05-2
Molecular Formula C5H6O5
Molecular Weight (g/mol) 146.098
MDL Number MFCD00002711
InChI Key OXTNCQMOKLOUAM-UHFFFAOYSA-N
Synonym 1,3-acetonedicarboxylic acid, 3-oxoglutaric acid, acetonedicarboxylic acid, pentanedioic acid, 3-oxo, acetone-1,3-dicarboxylic acid, 3-ketoglutaric acid, unii-ih7p7wo21p, 1,3-acetonedicarboxylic acid; 3-ketoglutaric acid, ih7p7wo21p, acetondicarbonsaure
PubChem CID 68328
IUPAC Name 3-oxopentanedioic acid
SMILES C(C(=O)CC(=O)O)C(=O)O

Specifications

Melting Point 126°C
Quantity 25 g
Formula Weight 146.10
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Acetonedicarboxylic Acid
EINECSNumber (2)-2762
TSCA Yes
Recommended Storage Freezer