missing translation for 'onlineSavingsMsg'
Learn More

1,3,6-Hexanetricarbonitrile 98.0+%, TCI America™

Catalog No. p-7131077
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15045G 5 g
H150425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15045G Supplier TCI America Supplier No. H15045G

Chemical Identifiers

CAS 1772-25-4
Molecular Formula C9H11N3
Molecular Weight (g/mol) 161.208
MDL Number MFCD00129792
InChI Key LNLFLMCWDHZINJ-UHFFFAOYSA-N
PubChem CID 15678
IUPAC Name hexane-1,3,6-tricarbonitrile
SMILES C(CC#N)CC(CCC#N)C#N

Specifications

Color Yellow
Boiling Point 200°C
Quantity 5 g
UN Number 3276
Formula Weight 161.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3,6-Hexanetricarbonitrile