accessibility menu, dialog, popup

UserName

NNN

CAS RN 1772-25-4

IUPAC Name: hexane-1,3,6-tricarbonitrile
Synonyms: hexane-1,3,6-tricarbonitrile
Molecular Weight (g/mol): 161.21
Molecular Formula: C9H11N3
InChi Key: LNLFLMCWDHZINJ-UHFFFAOYNA-N
SMILES: N#CCCCC(CCC#N)C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results