missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,6,8-Tetrabromocarbazole 95.0+%, TCI America™
$175.22 - $579.71
Chemical Identifiers
| CAS | 55119-09-0 |
|---|---|
| Molecular Formula | C12H5Br4N |
| Molecular Weight (g/mol) | 482.80 |
| MDL Number | MFCD00218272 |
| InChI Key | FNHISDQCWYSMTO-UHFFFAOYSA-N |
| PubChem CID | 258744 |
| IUPAC Name | 1,3,6,8-tetrabromo-9H-carbazole |
| SMILES | BrC1=CC2=C(NC3=C2C=C(Br)C=C3Br)C(Br)=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T2932200MG
|
TCI America
T2932200MG |
200 mg |
Each for $175.22
|
|
|||||
|
T29321G
|
TCI America
T29321G |
1 g |
Each for $579.71
|
|
|||||
Chemical Identifiers
| 55119-09-0 | |
| 482.80 | |
| FNHISDQCWYSMTO-UHFFFAOYSA-N | |
| 1,3,6,8-tetrabromo-9H-carbazole |
| C12H5Br4N | |
| MFCD00218272 | |
| 258744 | |
| BrC1=CC2=C(NC3=C2C=C(Br)C=C3Br)C(Br)=C1 |
Specifications
| 55119-09-0 | |
| White-Yellow | |
| MFCD00218272 | |
| FNHISDQCWYSMTO-UHFFFAOYSA-N | |
| 1,3,6,8-tetrabromo-9H-carbazole | |
| 258744 | |
| ≥95.0% (GC) | |
| 1,3,6,8-Tetrabromocarbazole |
| 229°C | |
| C12H5Br4N | |
| 200 mg | |
| BrC1=CC2=C(NC3=C2C=C(Br)C=C3Br)C(Br)=C1 | |
| 482.80 | |
| 482.80 | |
| Crystalline Powder |
Safety and Handling
TSCA : No