missing translation for 'onlineSavingsMsg'
Learn More

1,3,6,8-Pyrenetetrasulfonic acid tetrasodium salt, 95%

Catalog No. p-189180
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC325560010 1 g Glass bottle
AC325560050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC325560010 Supplier Thermo Scientific Chemicals Supplier No. 325560010
Only null left

CAS: 59572-10-0 | C16H6Na4O12S4 | 610.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59572-10-0
Molecular Formula C16H6Na4O12S4
Molecular Weight (g/mol) 610.44
MDL Number MFCD00042029
InChI Key UZBIRLJMURQVMX-UHFFFAOYSA-J
Synonym 1,3,6,8-pyrenetetrasulfonic acid, tetrasodium salt, unii-p062ld4v71, 1,3,6,8-pyrenetetrasulfonic acid tetrasodium salt, tetrasodium 1,3,6,8-pyrenetetrasulfonate, tetrasodium pyrene-1,3,6,8-tetrasulfonate, 1,3,6,8-pyrenetetrasulfonic acid, sodium salt 1:4, 1,3,6,8-pyrenetetrasulfonic acid tetrasodium salt hydrate, zlchem 1090, dsstox_cid_21233, dsstox_gsid_41233
PubChem CID 101082
IUPAC Name tetrasodium;pyrene-1,3,6,8-tetrasulfonate
SMILES C1=CC2=C3C(=C(C=C2S(=O)(=O)[O-])S(=O)(=O)[O-])C=CC4=C(C=C(C1=C43)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

CAS Min % 80.0
CAS Max % 100.0
Color Green
Infrared Spectrum Authentic
Assay Percent Range 85%
Packaging Glass bottle
Quantity 1 g
Formula Weight 610.44
Percent Purity 85%
Physical Form Fine Crystalline Powder
Chemical Name or Material 1,3,6,8-Pyrenetetrasulfonic acid tetrasodium salt, 85%
Show More Show Less

RUO – Research Use Only