missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Undecatriene (mixture of isomers) 75.0+%, TCI America™

Catalog No. p-7137861
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00625ML 5 mL
U006225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U00625ML Supplier TCI America Supplier No. U00625ML
Only null left

Chemical Identifiers

CAS 16356-11-9
Molecular Formula C11H18
Molecular Weight (g/mol) 150.265
MDL Number MFCD00015699
InChI Key JQQDKNVOSLONRS-JEGFTUTRSA-N
PubChem CID 5367412
IUPAC Name (3E,5E)-undeca-1,3,5-triene
SMILES CCCCCC=CC=CC=C

Specifications

Color Yellow
Boiling Point 88°C
Quantity 5 mL
Formula Weight 150.27
Percent Purity ≥75.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3,5-Undecatriene (mixture of isomers)
RTECSNumber YQ2833000
TSCA Yes
Recommended Storage Refrigerator