accessibility menu, dialog, popup

UserName

H3CCH2(E)(E)

CAS RN 16356-11-9

IUPAC Name: (3E,5E)-undeca-1,3,5-triene
Synonyms: undecatriene
Molecular Weight (g/mol): 150.27
Molecular Formula: C11H18
InChi Key: JQQDKNVOSLONRS-JEGFTUTRSA-N
SMILES: CCCCC\C=C\C=C\C=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results