missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tris(tert-butylthio)benzene 98.0+%, TCI America™

Catalog No. t22161g
Change view
Click to view available options
Quantity:
1 g
Catalog No. Quantity
T22161G 1 g
1 options

Catalog No. T22161G

Supplier: TCI America T22161G

Only null left

Chemical Identifiers

CAS 260968-03-4
Molecular Formula C18H30S3
Molecular Weight (g/mol) 342.618
InChI Key ZIHOGUOAVUUMKB-UHFFFAOYSA-N
PubChem CID 22617206
IUPAC Name 1,3,5-tris(tert-butylsulfanyl)benzene
SMILES CC(C)(C)SC1=CC(=CC(=C1)SC(C)(C)C)SC(C)(C)C

Specifications

Melting Point 133°C
Color White
Quantity 1 g
Formula Weight 342.62
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tris(tert-butylthio)benzene