accessibility menu, dialog, popup

UserName

CH3H3CCH3SSCH3H3CCH3SH3CCH3CH3

CAS RN 260968-03-4

IUPAC Name: 1,3,5-tris(tert-butylsulfanyl)benzene
Synonyms: 1,3,5-tris(tert-butylsulfanyl)benzene
Molecular Weight (g/mol): 342.62
Molecular Formula: C18H30S3
InChi Key: ZIHOGUOAVUUMKB-UHFFFAOYSA-N
SMILES: CC(C)(C)SC1=CC(SC(C)(C)C)=CC(SC(C)(C)C)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results