missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tris(chloromethyl)-2,4,6-trimethylbenzene, 97%

Catalog No. p-7056901
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66108MD 250 mg
AAH6610803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66108MD Supplier Thermo Scientific Chemicals Supplier No. H66108MD

CAS: 3849-01-2 | C12H15Cl3 | 265.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3849-01-2
Molecular Formula C12H15Cl3
Molecular Weight (g/mol) 265.60
MDL Number MFCD00229016
InChI Key PHQFMPNZCIHSPC-UHFFFAOYSA-N
IUPAC Name 1,3,5-tris(chloromethyl)-2,4,6-trimethylbenzene

Specifications

Color White
Quantity 250 mg
UN Number UN3261
Beilstein 2369523
Synonym 1,3,5-tris chloromethyl-2,4,6-trimethylbenzene, 1,3,5-tris-chloromethyl-2,4,6-trimethyl-benzene, benzene, 1,3,5-tris chloromethyl-2,4,6-trimethyl, ksc224a3n, tris-chloromethyl-mesitylene, 2,4,6-tris chloromethyl mesitylene, tris chloromethyl-2,4,6-trimethylbenzene, 1,3,5-trimethyl-2,4,6-tris chloromethyl benzene, benzene,3,5-tris chloromethyl-2,4,6-trimethyl, 1,3,5-tris chloromethyl-2,4,6-trimethyl-benzene
SMILES CC1=C(C(=C(C(=C1CCl)C)CCl)C)CCl
PubChem CID 291188
Formula Weight 265.6
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1,3,5-Tris(chloromethyl)-2,4,6-trimethylbenzene
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only