missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,5-Tris[4-(diphenylamino)phenyl]benzene 95.0+%, TCI America™
Supplier: TCI America T34361G
Specifications
| 1,3,5-Tris[4-(diphenylamino)phenyl]benzene | |
| White-Yellow | |
| MFCD30721946 | |
| XVMUGTFNHXHZIP-UHFFFAOYSA-N | |
| 3',5'-bis[4-(diphenylamino)phenyl]-N,N-diphenyl-[1,1'-biphenyl]-4-amine | |
| 16180236 | |
| ≥95.0% (N) |
| 147951-36-8 | |
| C60H45N3 | |
| 1 g | |
| C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 808.04 | |
| 808.04 | |
| Crystalline Powder |
Chemical Identifiers
| 147951-36-8 | |
| 808.04 | |
| XVMUGTFNHXHZIP-UHFFFAOYSA-N | |
| 3',5'-bis[4-(diphenylamino)phenyl]-N,N-diphenyl-[1,1'-biphenyl]-4-amine |
| C60H45N3 | |
| MFCD30721946 | |
| 16180236 | |
| C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1 |
Safety and Handling
TSCA : No