missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,5-Tris[4-(diphenylamino)phenyl]benzene 95.0+%, TCI America™
$233.00 - $691.57
Chemical Identifiers
| CAS | 147951-36-8 |
|---|---|
| Molecular Formula | C60H45N3 |
| Molecular Weight (g/mol) | 808.04 |
| MDL Number | MFCD30721946 |
| InChI Key | XVMUGTFNHXHZIP-UHFFFAOYSA-N |
| PubChem CID | 16180236 |
| IUPAC Name | 3',5'-bis[4-(diphenylamino)phenyl]-N,N-diphenyl-[1,1'-biphenyl]-4-amine |
| SMILES | C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T3436200MG
|
TCI America
T3436200MG |
200 mg |
Each for $233.00
|
|
|||||
|
T34361G
|
TCI America
T34361G |
1 g |
Each for $691.57
|
|
|||||
Chemical Identifiers
| 147951-36-8 | |
| 808.04 | |
| XVMUGTFNHXHZIP-UHFFFAOYSA-N | |
| 3',5'-bis[4-(diphenylamino)phenyl]-N,N-diphenyl-[1,1'-biphenyl]-4-amine |
| C60H45N3 | |
| MFCD30721946 | |
| 16180236 | |
| C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
| 147951-36-8 | |
| C60H45N3 | |
| 200 mg | |
| C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 808.04 | |
| 808.04 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD30721946 | |
| XVMUGTFNHXHZIP-UHFFFAOYSA-N | |
| 3',5'-bis[4-(diphenylamino)phenyl]-N,N-diphenyl-[1,1'-biphenyl]-4-amine | |
| 16180236 | |
| ≥95.0% (N) | |
| 1,3,5-Tris[4-(diphenylamino)phenyl]benzene |
Safety and Handling
TSCA : No