missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tris(4-aminophenyl)benzene 93.0+%, TCI America™

Catalog No. p-7137376
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27285G 5 g
T272825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27285G Supplier TCI America Supplier No. T27285G
Only null left

1,3,5-Tris(4-aminophenyl)benzene 93.0+%

Chemical Identifiers

CAS 118727-34-7
Molecular Formula C24H21N3
Molecular Weight (g/mol) 351.45
MDL Number MFCD18207723
InChI Key QHQSCKLPDVSEBJ-UHFFFAOYSA-N
PubChem CID 12058813
IUPAC Name 3',5'-bis(4-aminophenyl)-[1,1'-biphenyl]-4-amine
SMILES NC1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=C(N)C=C1)C1=CC=C(N)C=C1

Specifications

Melting Point 165°C
Quantity 5 g
Formula Weight 351.45
Percent Purity ≥93.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tris(4-aminophenyl)benzene