accessibility menu, dialog, popup

UserName

NH2H2NNH2

CAS RN 118727-34-7

IUPAC Name: 3',5'-bis(4-aminophenyl)-[1,1'-biphenyl]-4-amine
Synonyms: 3',5'-bis(4-aminophenyl)-[1,1'-biphenyl]-4-amine
Molecular Weight (g/mol): 351.45
Molecular Formula: C24H21N3
InChi Key: QHQSCKLPDVSEBJ-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=C(N)C=C1)C1=CC=C(N)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results