missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,5-Tris(2-propynyloxy)benzene 98.0+%, TCI America™
$380.27 - $1314.16
Chemical Identifiers
| CAS | 114233-80-6 |
|---|---|
| Molecular Formula | C15H12O3 |
| Molecular Weight (g/mol) | 240.258 |
| InChI Key | WVZIFQJSKQJHDU-UHFFFAOYSA-N |
| Synonym | 1,3,5-Tris(propargyloxy)benzene |
| PubChem CID | 11630083 |
| IUPAC Name | 1,3,5-tris(prop-2-ynoxy)benzene |
| SMILES | C#CCOC1=CC(=CC(=C1)OCC#C)OCC#C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T3135200MG
|
TCI America
T3135200MG |
200 mg |
Each for $380.27
|
|
|||||
|
T31351G
|
TCI America
T31351G |
1 g |
Each for $1,314.16
|
|
|||||
Chemical Identifiers
| 114233-80-6 | |
| 240.258 | |
| 1,3,5-Tris(propargyloxy)benzene | |
| 1,3,5-tris(prop-2-ynoxy)benzene |
| C15H12O3 | |
| WVZIFQJSKQJHDU-UHFFFAOYSA-N | |
| 11630083 | |
| C#CCOC1=CC(=CC(=C1)OCC#C)OCC#C |
Specifications
| 114233-80-6 | |
| White-Yellow | |
| 200 mg | |
| WVZIFQJSKQJHDU-UHFFFAOYSA-N | |
| 1,3,5-tris(prop-2-ynoxy)benzene | |
| 11630083 | |
| ≥98.0% (T) | |
| 1,3,5-Tris(2-propynyloxy)benzene |
| 82°C | |
| C15H12O3 | |
| 1,3,5-Tris(propargyloxy)benzene | |
| C#CCOC1=CC(=CC(=C1)OCC#C)OCC#C | |
| 240.258 | |
| 240.26 | |
| Crystalline Powder |
Safety and Handling
TSCA : No