missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tris(2-hydroxyethyl)cyanuric acid, 97%

Catalog No. p-7025952
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2169718 50 g
AAB2169730 250 g
AAB216970B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2169718 Supplier Thermo Scientific Chemicals Supplier No. B2169718
Only null left

CAS: 839-90-7 | C9H15N3O6 | 261.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 839-90-7
Molecular Formula C9H15N3O6
Molecular Weight (g/mol) 261.23
MDL Number MFCD00003549
InChI Key BPXVHIRIPLPOPT-UHFFFAOYSA-N
Synonym 1,3,5-tris 2-hydroxyethyl cyanuric acid, theic, tris 2-hydroxyethyl isocyanurate, tris hydroxyethyl cyanurate, tris 2-hydroxyethyl cyanurate, tris hydroxyethyl isocyanurate, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3,5-tris 2-hydroxyethyl, 1,3,5-tris 2-hydroxyethyl-1,3,5-triazinane-2,4,6-trione, unii-n7w2mh0p2m
PubChem CID 13286
SMILES OCCN1C(=O)N(CCO)C(=O)N(CCO)C1=O

Specifications

Melting Point 136°C to 140°C
Quantity 50 g
IUPAC Name tris(2-hydroxyethyl)-1,3,5-triazinane-2,4,6-trione
Formula Weight 261.24
Percent Purity 97%
Chemical Name or Material 1,3,5-Tris(2-hydroxyethyl)cyanuric acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 212-660-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only