missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triphenylbenzene, 99+%

CAS: 612-71-5 | C24H18 | 306.41 g/mol

$99.74 - $1341.63

Chemical Identifiers

CAS 612-71-5
Molecular Formula C24H18
Molecular Weight (g/mol) 306.41
MDL Number MFCD00003060
InChI Key SXWIAEOZZQADEY-UHFFFAOYSA-N
Synonym triphenylbenzene, 5'-phenyl-1,1':3',1-terphenyl, s-triphenylbenzene, m-terphenyl, 5'-phenyl, 1,1':3',1-terphenyl, 5'-phenyl, 5'-phenyl-m-terphenyl, benzene, 1,3,5-triphenyl, 1,1'-biphenyl, 3,5-diphenyl, unii-408mso5wo3, 3,5-diphenyl-1,1'-biphenyl
PubChem CID 11930
IUPAC Name 1,3,5-triphenylbenzene
SMILES C1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2135318
View Documents
Thermo Scientific Chemicals
B2135318
50 g
Each for $99.74
Only null left
 
AAB2135330
View Documents
Thermo Scientific Chemicals
B2135330
250 g
Each for $406.01
Only null left
 
AAB213530B
View Documents
Thermo Scientific Chemicals
B213530B
1000 g
Each for $1,341.63
Only null left
 
Description

Description

1,3,5-Triphenylbenzene fluorophore as a selective Cu2+ sensor in aqueous media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Triphenylbenzene fluorophore as a selective Cu2+ sensor in aqueous media.

Solubility
Solubility in toluene is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

612-71-5
306.41
SXWIAEOZZQADEY-UHFFFAOYSA-N
11930
C1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1
C24H18
MFCD00003060
triphenylbenzene, 5'-phenyl-1,1':3',1-terphenyl, s-triphenylbenzene, m-terphenyl, 5'-phenyl, 1,1':3',1-terphenyl, 5'-phenyl, 5'-phenyl-m-terphenyl, benzene, 1,3,5-triphenyl, 1,1'-biphenyl, 3,5-diphenyl, unii-408mso5wo3, 3,5-diphenyl-1,1'-biphenyl
1,3,5-triphenylbenzene

Specifications

612-71-5
460°C
MFCD00003060
triphenylbenzene, 5'-phenyl-1,1':3',1-terphenyl, s-triphenylbenzene, m-terphenyl, 5'-phenyl, 1,1':3',1-terphenyl, 5'-phenyl, 5'-phenyl-m-terphenyl, benzene, 1,3,5-triphenyl, 1,1'-biphenyl, 3,5-diphenyl, unii-408mso5wo3, 3,5-diphenyl-1,1'-biphenyl
C1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1
306.41
306.41
1,3,5-Triphenylbenzene
172°C to 174°C
C24H18
50 g
SXWIAEOZZQADEY-UHFFFAOYSA-N
1,3,5-triphenylbenzene
11930
≥99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 210-318-3

RTECSNumber : WZ6530000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only