accessibility menu, dialog, popup

UserName

CAS RN 612-71-5

IUPAC Name: 3,5-diphenyl-1,1'-biphenyl
Synonyms: 3,5-diphenyl-1,1'-biphenyl
Molecular Weight (g/mol): 306.41
Molecular Formula: C24H18
InChi Key: SXWIAEOZZQADEY-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)