missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triphenylbenzene, 99+%

Catalog No. AAB2135318
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2135318 50 g
AAB2135330 250 g
AAB213530B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2135318 Supplier Thermo Scientific Chemicals Supplier No. B2135318
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 612-71-5 | C24H18 | 306.41 g/mol

1,3,5-Triphenylbenzene fluorophore as a selective Cu2+ sensor in aqueous media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Triphenylbenzene fluorophore as a selective Cu2+ sensor in aqueous media.

Solubility
Solubility in toluene is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 612-71-5
Molecular Formula C24H18
Molecular Weight (g/mol) 306.41
MDL Number MFCD00003060
InChI Key SXWIAEOZZQADEY-UHFFFAOYSA-N
Synonym triphenylbenzene, 5'-phenyl-1,1':3',1-terphenyl, s-triphenylbenzene, m-terphenyl, 5'-phenyl, 1,1':3',1-terphenyl, 5'-phenyl, 5'-phenyl-m-terphenyl, benzene, 1,3,5-triphenyl, 1,1'-biphenyl, 3,5-diphenyl, unii-408mso5wo3, 3,5-diphenyl-1,1'-biphenyl
PubChem CID 11930
IUPAC Name 1,3,5-triphenylbenzene
SMILES C1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 172°C to 174°C
Boiling Point 460°C
Quantity 50 g
Formula Weight 306.41
Percent Purity ≥99%
Chemical Name or Material 1,3,5-Triphenylbenzene
EINECSNumber 210-318-3
RTECSNumber WZ6530000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only