missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trinitrobenzene (wetted with ca. 40% Water) (unit weight on dry weight basis) 98.0+%, TCI America™

Catalog No. t049610g
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T049610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T049610G Supplier TCI America Supplier No. T049610G
Only null left

Chemical Identifiers

CAS 99-35-4
Molecular Formula C6H3N3O6
Molecular Weight (g/mol) 213.105
MDL Number MFCD00059179
InChI Key UATJOMSPNYCXIX-UHFFFAOYSA-N
Synonym trinitrobenzene, sym-trinitrobenzene, s-trinitrobenzene, benzite, syn-trinitrobenzene, trinitrobenzeen, trinitrobenzol, benzene, 1,3,5-trinitro, benzene, trinitro-wet, rcra waste number u234
PubChem CID 7434
ChEBI CHEBI:48113
IUPAC Name 1,3,5-trinitrobenzene
SMILES C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 122°C
Color Yellow
Quantity 10 g
UN Number 1354
Formula Weight 213.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Trinitrobenzene (wetted with ca. 40% Water) (unit weight on dry weight basis)
EINECSNumber (3)-2753
RTECSNumber DC3850000
TSCA Yes